2-Methylheptane 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Isoheptane
- CAS Number:592-27-8
- Formula:C₈H₁₈
- MDL Number:MFCD00009518
- Molecular Weight:114.23 g/mol
- Boiling Pt:116 °C (761 mmHg)
- Melting Pt:–108.9 °C
- Density:0.69 g/cm³
- Flash Pt:4 °C
- Storage Temperature:Ambient
- EINECS:209-747-9
- UN:1262
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3930 to 1.3970 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



