2-Methylheptane 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Isoheptane
- CAS Number:592-27-8
- Formula:C₈H₁₈
- MDL Number:MFCD00009518
- Molecular Weight:114.23 g/mol
- Boiling Pt:116 °C (761 mmHg)
- Melting Pt:–108.9 °C
- Density:0.69 g/cm³
- Flash Pt:4 °C
- Storage Temperature:Ambient
- EINECS:209-747-9
- UN:1262
- ADR:3,II
Documents
ACRO12688
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3930 to 1.3970 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



