Methylcyclohexane 99%, extra pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Methylcyclohexane 99%, extra pure
Catalog # ACRO126580010
Supplier: Thermo Fisher Scientific
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126.3 °C
- Density:0.77 g/cm³
- Flash Pt:–3 °C
- Storage Temperature:Ambient
- EINECS:203-624-3
- UN:2296
- ADR:3,II
Documents
ACRO12658
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4240 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.01 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



