Vanillic acid 97+%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Hydroxy-3-methoxybenzoic acid
- CAS Number:121-34-6
- Formula:C₈H₈O₄
- MDL Number:MFCD00002551
- Molecular Weight:168.15 g/mol
- Merck Index:12,10068
- Boiling Pt:353 °C (1013 hPa)
- Melting Pt:210…214 °C
- Storage Temperature:Ambient
- EINECS:204-466-8
Documents
ACRO12135
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:208 to 214 °C
- HPLC:≥97.0 %
- GC:≥99.0 %
- Color scale:≤50 APHA
- Free acid:≤0.5 % (as Crotonic acid)
- Suitability:(for use with Dithizone) Passes test
- Substance darkened by H2SO4:≤5 APHA
- Free chlorine:≤5 ppm
- Calcium (Ca):≤0.1 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.2 ppm
- Zinc (Zn):≤0.05 ppm
- Impurity:≤0.005 % Carbon tetrachloride
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.5 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



