Hydroquinone 99%
Plastic bottle, 2 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285…287 °C (760 mmHg)
- Melting Pt:170…174 °C
- Density:1,332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Documents
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:170 to 175 °C
- HPLC:≥98.5 %
Specifications
- Pack type:Plastic bottle
- Pk:2 kg



