Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Hexadecanol 96%

1-Hexadecanol 96%

Plastic bottle, 1 kg
Catalog # ACRO120480010
Irritant
1-Hexadecanol 96%
Catalog # ACRO120480010
CAS Number:  36653-82-4
1-Hexadecanol 96%
1-Hexadecanol 96%

Plastic bottle, 1 kg

CAS Number:  36653-82-4
Price
447,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cetyl alcohol, Palmityl alcohol
  • CAS Number:36653-82-4
  • Formula:H₃C(CH₂)₁₄CH₂OH
  • MDL Number:MFCD00004760
  • Molecular Weight:242.45 g/mol
  • Merck Index:13,02037
  • Boiling Pt:344 °C (760 mmHg)
  • Melting Pt:47…50 °C
  • Density:0,81 g/cm³ (20 °C)
  • Flash Pt:>157 °C
  • Storage Temperature:Ambient
  • EINECS:253-149-0

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder or flakes or pellets
  • Infrared spectrum:
    Conforms
  • GC:
    ≥95 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.