Eugenol 99%
Glass bottle for solids, 500 g
: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- :97-53-0
- :C₁₀H₁₂O₂
- :MFCD00008654
- :164.2 g/mol
- :13,03931
- :254 °C (760 mmHg)
- :12…10 °C
- :1.06 g/cm³
- :>112 °C
- :Ambient
- :202-589-1
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5400 to 1.5420 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(20°C) 1.064 to 1.070
- Pack type:Glass bottle for solids
- Pk:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



