Eugenol 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
- CAS Number:97-53-0
- Formula:C₁₀H₁₂O₂
- MDL Number:MFCD00008654
- Molecular Weight:164.2 g/mol
- Merck Index:13,03931
- Boiling Pt:254 °C (760 mmHg)
- Melting Pt:12…10 °C
- Density:1.06 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:202-589-1
Documents
ACRO11911
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5400 to 1.5420 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(20°C) 1.064 to 1.070
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



