Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Eugenol 99%

Glass bottle for solids, 5 g
Katalog # ACRO119110050
Irritant
Leverandør:   Thermo Fisher Scientific
Eugenol 99%
Katalog # ACRO119110050
CAS nummer:  97-53-0
Eugenol 99%
Eugenol 99%

Glass bottle for solids, 5 g

CAS nummer:  97-53-0
Price
347,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole
  • CAS Number:97-53-0
  • Formula:C₁₀H₁₂O₂
  • MDL Number:MFCD00008654
  • Molecular Weight:164.2 g/mol
  • Merck Index:13,03931
  • Boiling Pt:254 °C (760 mmHg)
  • Melting Pt:12…10 °C
  • Density:1.06 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Ambient
  • EINECS:202-589-1

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-green Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5400 to 1.5420 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific gravity:
    (20°C) 1.064 to 1.070

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications