2-Thiobarbituric acid 98%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
2-Thiobarbituric acid 98%
Catalog # ACRO114965000
Supplier: Thermo Fisher Scientific
CAS Number: 504-17-6
Jump to:
Product Details & Documents
Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine
- CAS Number:504-17-6
- Formula:C₄H₄N₂O₂S
- MDL Number:MFCD00006674
- Molecular Weight:144.15 g/mol
- Boiling Pt:419 °C (1013 hPa)
- Melting Pt:240…255 °C
- Density:1,661 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-985-8
Documents
ACRO11496
SDS
Specifications
Specification Test Results
- Appearance:White to yellow to light brown Powder or crystalline solid
- Infrared spectrum:Conforms
- Solubility:(5% in 1M NaOH) Clear to slightly hazy
- HPLC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



