2-Thiobarbituric acid 98%
Plastic bottle, 100 g
: 4,6-Dihydroxy-2-mercaptopyrimidine
- :504-17-6
- :C₄H₄N₂O₂S
- :MFCD00006674
- :144.15 g/mol
- :419 °C (1013 hPa)
- :240…255 °C
- :1,661 g/cm³ (20 °C)
- :Ambient
- :207-985-8
Documents
ACRO11496
SDS
- Appearance:White to yellow to light brown Powder or crystalline solid
- Infrared spectrum:Conforms
- Solubility:(5% in 1M NaOH) Clear to slightly hazy
- HPLC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
- Pack type:Plastic bottle
- Pk:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



