Imidazole, Millipore®
Glass bottle for solids, 100 g
Catalog # 4015-100
Supplier: Merck Millipore (Calbiochem)
Imidazole, Millipore®
Catalog # 4015-100
Supplier: Merck Millipore (Calbiochem)
CAS Number: 288-32-4
Jump to:
Product Details & Documents
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
- CAS Number:288-32-4
- Formula:C₃H₄N₂
- MDL Number:MFCD00005183
- Molecular Weight:68.08 g/mol
- Merck Index:13,04935
- Boiling Pt:255 °C (1013 hPa)
- Melting Pt:89…90 °C
- Density:1.18 g/cm³ (20 °C)
- Flash Pt:135 °C
- Storage Temperature:Ambient
- EINECS:206-019-2
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



