Cyclohexane 99+%, pure
Plastic drum, 25 L
Supplier: Thermo Fisher Scientific
Cyclohexane 99+%, pure
Catalog # ACRO111110250
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:6.5 °C
- Density:0.77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
ACRO11111
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (K.F.)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
Specifications
- Pack type:Plastic drum
- Pk:25 L



