Aluminon
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
- CAS Number:569-58-4
- Formula:C₂₂H₂₃N₃O₉
- MDL Number:MFCD00040925
- Molecular Weight:473.44 g/mol
- Melting Pt:220…225 °C (dec.)
- Storage Temperature:Ambient
- EINECS:209-319-1
Documents
ACRO10306
SDS
Specifications
Specification Test Results
- Appearance:Red to yellow-brown to red-brown Crystalline powder
- Infrared spectrum:Conforms
- Sulfated ash:≤0.2 %
- Solubility:(1.0 % in water) Clear
- Suitability:(for Al determination) Passes test
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



