Aluminon
Glass bottle, 25 g
: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
- :569-58-4
- :C₂₂H₂₃N₃O₉
- :MFCD00040925
- :473.44 g/mol
- :220…225 °C (dec.)
- :Ambient
- :209-319-1
- Appearance:Red to yellow-brown to red-brown Crystalline powder
- Infrared spectrum:Conforms
- Sulfated ash:≤0.2 %
- Solubility:(1.0 % in water) Clear
- Suitability:(for Al determination) Passes test
- Pack type:Glass bottle
- Pk:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



