Hose clips
20 - 22 mm, White
Catalog # BURK8678-0022
Supplier: BURKLE
Jump to:
Product Details & Documents
Hose clips made of Polyacetal (POM).
Specifications
- Spanning width:20 - 22 mm
- Colour:White
- Seq:10