Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hose clips
Hose clips
13 - 15 mm, White
Catalog # BURK8678-0015
Supplier:   BURKLE
Hose clips
Catalog # BURK8678-0015
Supplier:  BURKLE
Hose clips
Hose clips
13 - 15 mm, White
Price
46,30 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Hose clips made of Polyacetal (POM).
Specifications
  • Spanning width:
    13 - 15 mm
  • Colour:
    White
  • Seq:
    06