Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hose clips
Hose clips
11 - 12 mm, White
Catalog # 229-0261
Supplier:   BURKLE
Hose clips
Catalog # 229-0261
Supplier:  BURKLE
Hose clips
Hose clips
11 - 12 mm, White
Price
46,30 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Hose clips made of Polyacetal (POM).
Specifications
  • Spanning width:
    11 - 12 mm
  • Colour:
    White
  • Seq:
    04