Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
kr0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Clamps
Tubing and Sanitary Clamps
Hose clips
Hose clips
Print
Hose clips
10 - 11 mm, White
Catalog # BURK8678-0011
Supplier:
BURKLE
Hose clips
Catalog # BURK8678-0011
Supplier:
BURKLE
Expand image
Return to Top
Hose clips
10 - 11 mm, White
Specifications
Add to cart
Jump to:
Specifications
Price
46,30 kr
Log in
for availability
Unit of Measure
1 * 10 Items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Hose clips made of Polyacetal (POM).
Specifications
Spanning width:
10 - 11 mm
Colour:
White
Seq:
03