D-(-)-Mannitol ACS
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290 - 295 °C (4 hPa)
- Melting Pt:165…170 °C
- Density:1,49 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-711-8
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.05 % (5 g, 105°C, 2 hrs)
- Residue after ignition:≤0.01 %
- Specific optical rotation:+23.3° to +24.3° (25°C, 589 nm)
- Heavy metals (ICP-OES):≤5 ppm (as Pb)
- Titrable acid:≤0.0008 meq/g
- Insoluble matter:≤0.01 %
- Reducing sugars:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



