Tidy tray
12, 402×302×60 mm
Catalog # BRND768510
Supplier: Brand
Jump to:
Product Details & Documents
PVC, white.
- Version with 12 compartments for small items, e.g. magnetic stirrer bars
- Version with five compartments for small parts of various sizes, e.g. pipettes, thermometers, test tubes, spatulas, etc.
- Smooth surfaces with rounded corners for easy cleaning
For well organised storage of many items in drawers, on shelves or at the workstation.
Specifications
- Compartments:12
- W×D×H:402×302×60 mm