Silver nitrate 99.5% for analysis
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Silver nitrate 99.5% for analysis
Catalog # ACRO419365000
Supplier: Thermo Fisher Scientific
CAS Number: 7761-88-8
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (1013 hPa)
- Melting Pt:212 °C
- Density:4,35 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Documents
ACRO41936
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99.4 %
- Chloride (Cl):≤5 ppm
- Cadmium (Cd):≤1 ppm
- Calcium (Ca):≤10 ppm
- Chromium (Cr):≤0.2 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.4 ppm
- Lead (Pb):≤0.2 ppm
- Manganese (Mn):≤0.1 ppm
- Nickel (Ni):≤0.2 ppm
- Palladium (Pd):≤0.1 ppm
- Sulfur (S):≤20 ppm
- Zinc (Zn):≤10 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



