Order Entry
Denmark
Orders LinkContactUsLinkComponent
Silver nitrate 99.5% for analysis

Silver nitrate 99.5% for analysis

Glass bottle for solids, 100 g
Catalog # ACRO419361000
Oxidizing
Corrosive
Environment
Silver nitrate 99.5% for analysis
Catalog # ACRO419361000
CAS Number:  7761-88-8
Silver nitrate 99.5% for analysis
Silver nitrate 99.5% for analysis

Glass bottle for solids, 100 g

CAS Number:  7761-88-8
Price
3.530,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:212 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Argentometric:
    ≥99.4 %
  • Chloride (Cl):
    ≤5 ppm
  • Cadmium (Cd):
    ≤1 ppm
  • Calcium (Ca):
    ≤10 ppm
  • Chromium (Cr):
    ≤0.2 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.4 ppm
  • Lead (Pb):
    ≤0.2 ppm
  • Manganese (Mn):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.2 ppm
  • Palladium (Pd):
    ≤0.1 ppm
  • Sulfur (S):
    ≤20 ppm
  • Zinc (Zn):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.