Pyrogallol ACS
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
Documents
ACRO41862
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131.0 to 135.0 °C
- Residue after ignition:≤0.005 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:500 g



