Pyrogallol ACS
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS Number:87-66-1
- Formula:C₆H₆O₃
- MDL Number:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Boiling Pt:309 °C (1013 hPa)
- Melting Pt:131…135 °C
- Density:1,453 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-762-9
- UN:2811
- ADR:6.1,III
Documents
ACRO41862
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131.0 to 135.0 °C
- Residue after ignition:≤0.005 %
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤50 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



