Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Papain

Glass bottle, 500 g
Catalog # ACRO416765000
Papain
Catalog # ACRO416765000
CAS Number:  
Papain
Papain

Glass bottle, 500 g

CAS Number:  
Price
5.590,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Cat. No.:
    ACRO416765000
  • Form:
    Cream to light brown Powder
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131791
  • Enzyme name:
    Papain
  • CAS No.:
    9001-73-4

Specifications