Order Entry
Denmark
Orders LinkContactUsLinkComponent
Papain

Papain

Glass bottle, 100 g
Catalog # ACRO416761000
Papain
Catalog # ACRO416761000
CAS Number:  
Papain
Papain

Glass bottle, 100 g

CAS Number:  
Price
1.470,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 9001-73-4

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Cat. No.:
    ACRO416761000
  • Form:
    Cream to light brown Powder
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131791
  • Enzyme name:
    Papain
  • CAS No.:
    9001-73-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.