Order Entry
Denmark
Orders LinkContactUsLinkComponent
Papain

Papain

Glass bottle, 10 g
Catalog # ACRO416760100
Papain
Catalog # ACRO416760100
CAS Number:  
Papain
Papain

Glass bottle, 10 g

CAS Number:  
Price
305,00 kr
Log in for availability
1 * 10 g
Product Details & Documents

CAS No.: 9001-73-4

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Cat. No.:
    ACRO416760100
  • Form:
    Cream to light brown Powder
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00131791
  • Enzyme name:
    Papain
  • CAS No.:
    9001-73-4

Specifications