Order Entry
Denmark
Orders LinkContactUsLinkComponent
Ninhydrin ACS

Ninhydrin ACS

Glass bottle for solids, 10 g
Catalog # ACRO415720100
Irritant
Ninhydrin ACS
Catalog # ACRO415720100
CAS Number:  485-47-2
Ninhydrin ACS
Ninhydrin ACS

Glass bottle for solids, 10 g

CAS Number:  485-47-2
Price
412,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250 °C (dec.)
  • Density:0,862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light brown Crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    Passes test
  • Solubility:
    Passes test
  • Identification:
    Passes test
  • Sensitivity:
    (to amino acids) Passes test

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.