Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bulb pipettes (one mark), BLAUBRAND®, class AS

Bulb pipettes (one mark), BLAUBRAND®, class AS

10 ml, 0,02 ± ml, 440 mm
Catalog # 612-1212
Supplier:   Brand
Bulb pipettes (one mark), BLAUBRAND®, class AS
Catalog # 612-1212
Supplier:  Brand
Bulb pipettes (one mark), BLAUBRAND®, class AS
Bulb pipettes (one mark), BLAUBRAND®, class AS

10 ml, 0,02 ± ml, 440 mm

Price
82,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time: 5 seconds
  • Batch certificate
  • Also available (on request) with an individual certificate or DAkkS calibration certificate
  • USP version available

DIN EN ISO 648

Certifications: DIN EN ISO 648; DE-M marking.

Specifications
  • Capacity:
    10 ml
  • Length:
    440 mm
  • Seq:
    13
  • Tolerance:
    0,02 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.