Watch glasses
Watch glass, PTFE, 80 mm
Catalog # BRND150912
Supplier: Brand
Jump to:
Product Details & Documents
PTFE, white.
- Very good chemical resistance
- Temperature resistant from –200 to +280 °C
- Exceptionally smooth surfaces to prevent contamination
Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.
Specifications
- Ø:80 mm
- Description:Watch glass, PTFE
- Seq:07
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



