Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stoppers, rubber

Stoppers, rubber

30,0 mm, 21,0 mm, 27,0 mm
Catalog # BRND144390
Supplier:   Brand
Stoppers, rubber
Catalog # BRND144390
Supplier:  Brand
Stoppers, rubber
Stoppers, rubber

30,0 mm, 21,0 mm, 27,0 mm

Price
485,00 kr
Log in for availability
1 * 25 Items
Product Details & Documents

Natural rubber, red.

Specifications
  • Ø bottom:
    21,0 mm
  • Ø top:
    27,0 mm
  • Height:
    30,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.