Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

5000 ml, 210 mm, 180 mm
Katalog # BOCH8506
Leverandør:   BOCHEM
Beakers, low form
Katalog # BOCH8506
Leverandør:  BOCHEM
Beakers, low form
Beakers, low form

5000 ml, 210 mm, 180 mm

Price
2.450,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    180 mm
  • Capacity:
    5000 ml
  • Height:
    210 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.