Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clamps
Clamps
100 mm
Catalog # 241-0401
Supplier:   BOCHEM
Clamps
Catalog # 241-0401
Supplier:  BOCHEM
Clamps
Clamps
100 mm
Price
588,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifikationer
  • Spanning width:
    100 mm