Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double bossheads
Thumb screw, Aluminium, 16,5 mm
  241-7204
 :   BOCHEM
Double bossheads
  241-7204
 :  BOCHEM
Double bossheads
Double bossheads
Thumb screw, Aluminium, 16,5 mm
87,20 kr
1 * 1 Items
For stand assembly.
  • Quick, easy assembly and disassembly
  • Robust, durable
  • Description:
    Thumb screw
  • Material:
    Aluminium
  • Seq:
    1
  • For rod Ø:
    16,5 mm