Order Entry
Denmark
Orders LinkContactUsLinkComponent
Double bossheads
Double bossheads
Heart screw, Stainless steel, 16,5 mm
Catalog # 241-7221
Supplier:   BOCHEM
Double bossheads
Catalog # 241-7221
Supplier:  BOCHEM
Double bossheads
Double bossheads
Heart screw, Stainless steel, 16,5 mm
Price
1.010,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
For stand assembly.
  • Quick, easy assembly and disassembly
  • Robust, durable
Specifications
  • Description:
    Heart screw
  • Material:
    Stainless steel
  • For rod Ø:
    16,5 mm