Order Entry
Denmark
Orders LinkContactUsLinkComponent
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
24/29 NS, 24/29 NS
Catalog # 271-0016
Supplier:   SciLabware
Adapters, receiver, plain bent, Quickfit®
Catalog # 271-0016
Supplier:  SciLabware
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
24/29 NS, 24/29 NS
Price
406,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Receiver, delivery, PYREX® borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Seq:
    04