Order Entry
Denmark
Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 150 mm
Catalog # 216-2777
Supplier:   SciLabware
Watch glasses
Catalog # 216-2777
Supplier:  SciLabware
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 150 mm
Price
254,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
  • Excellent chemical resistance
  • High temperature resistance
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifications
  • Ø:
    150 mm
  • Description:
    Watch glass, PYREX® borosilicate glass with firepolished edges
  • Seq:
    09