Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(+)-Biotin 98%

D-(+)-Biotin 98%

Glass bottle for solids, 1 g
Catalog # ACRO230090010
D-(+)-Biotin 98%
Catalog # ACRO230090010
CAS Number:  58-85-5
D-(+)-Biotin 98%
D-(+)-Biotin 98%

Glass bottle for solids, 1 g

CAS Number:  58-85-5
Price
1.220,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:231…233 °C (dec.)
  • Density:1,41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Sulfated ash:
    ≤0.1 %
  • Appearance of solution:
    (2% soln. in 0.1 N NaOH) clear, colorless
  • Specific optical rotation:
    +89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH) (on dried substance)
  • Titration with NaOH:
    ≥97.5 % (On dried substance)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.