Order Entry
Denmark
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 5 g
Catalog # ACRO229280050
Irritant
Health
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
Catalog # ACRO229280050
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 5 g

CAS Number:  54827-17-7
Price
2.430,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:167…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    167 to 172 °C
  • Loss on drying:
    ≤0.5 %
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥99.0 %
  • Titration with HClO4:
    98.0 to 102.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.