D-(-)-Serine 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:220 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-229-4
Documents
ACRO22707
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder or crystals
- Infrared spectrum:Conforms
- Loss on drying:≤0.3 % (105°C, 3 h)
- Sulfated ash:≤0.1 %
- Specific optical rotation:-13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with HClO4:97.5 to 102.5 %
- Other amino acids:≤0.5 % (TLC)
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



