Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Serine 98%

D-(-)-Serine 98%

Glass bottle for solids, 5 g
Catalog # ACRO227070050
D-(-)-Serine 98%
Catalog # ACRO227070050
CAS Number:  312-84-5
D-(-)-Serine 98%
D-(-)-Serine 98%

Glass bottle for solids, 5 g

CAS Number:  312-84-5
Price
284,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (R)-(-)-Serine, H-D-Ser-OH
  • CAS Number:312-84-5
  • Formula:C₃H₇NO₃
  • MDL Number:MFCD00004269
  • Molecular Weight:105.09 g/mol
  • Merck Index:12,08604
  • Melting Pt:220 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-229-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.3 % (105°C, 3 h)
  • Sulfated ash:
    ≤0.1 %
  • Specific optical rotation:
    -13.0° to -16.0° (20°C, 589 nm) (c=10, 2 N HCl)
  • Titration with HClO4:
    97.5 to 102.5 %
  • Other amino acids:
    ≤0.5 % (TLC)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications