Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Strap clamp for securing vessels, stand Ø 8 - 16 mm, vessel Ø 40 - 300 mm, T 18 basic, T 25 digital, Clamp device RH 3
Catalog # 431-2943
Supplier:   IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Catalog # 431-2943
Supplier:  IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Strap clamp for securing vessels, stand Ø 8 - 16 mm, vessel Ø 40 - 300 mm, T 18 basic, T 25 digital, Clamp device RH 3

Price
2.900,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Strap clamp for securing vessels, stand Ø 8 - 16 mm, vessel Ø 40 - 300 mm
  • Seq:
    103
  • Type:
    Clamp device RH 3
  • For:
    T 18 basic, T 25 digital
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    431-2944
    Log in for availability
    1 * 1 Items - 3.570,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.