Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Bosshead clamp, 6 - 16 mm Ø stand, 6 - 16 mm Ø extension arm, T 18 basic, T 25 digital, Fixing clamp R 182
Catalog # 431-2923
Supplier:   IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Catalog # 431-2923
Supplier:  IKA
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic
Accessories for T 10 basic, T 18 basic, T 25 digital and T 50 basic

Bosshead clamp, 6 - 16 mm Ø stand, 6 - 16 mm Ø extension arm, T 18 basic, T 25 digital, Fixing clamp R 182

Price
902,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Bosshead clamp, 6 - 16 mm Ø stand, 6 - 16 mm Ø extension arm
  • Seq:
    101
  • Type:
    Fixing clamp R 182
  • For:
    T 18 basic, T 25 digital
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm
    431-2944
    Log in for availability
    1 * 1 Items - 3.570,00 kr

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.