Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bulb pipettes, one mark, class B

Bulb pipettes, one mark, class B

50 ml, 0,075 ± ml, 540 mm, -
Catalog # 612-1357
Supplier:   HIRSCHMANN
Bulb pipettes, one mark, class B
Catalog # 612-1357
Supplier:  HIRSCHMANN
Bulb pipettes, one mark, class B
Bulb pipettes, one mark, class B

50 ml, 0,075 ± ml, 540 mm, -

Price
99,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

AR-Glas®

  • Calibrated to deliver (TD, Ex)
  • With silver diffusion ink durable brown graduations

DIN 12690

 : DIN EN ISO 648; DE-M marking

Specifications
  • Capacity:
    50 ml
  • Length:
    540 mm
  • Suppl. to DIN:
    -
  • Tolerance:
    0,075 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.