Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tools

Tools

Open end wrench, 12 mm, 116 mm long
Katalog # HEWL8710-1841
Leverandør:   AGILENT HEWLETT PACKARD
Tools
Katalog # HEWL8710-1841
Tools
Tools

Open end wrench, 12 mm, 116 mm long

Price
194,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Description:
    Open end wrench, 12 mm, 116 mm long
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.