Certified microvolume inserts for wide opening vials
Inserts, deactivated, 0,40 ml, Flat bottom, Glass, No
Catalog # HEWL5183-2086
Supplier: AGILENT HEWLETT PACKARD
Certified microvolume inserts for wide opening vials
Catalog # HEWL5183-2086
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:0,40 ml
- Description:Inserts, deactivated
- Material:Glass
- Seq:05
- Base:Flat bottom