Order Entry
Denmark
Orders LinkContactUsLinkComponent
Certified microvolume inserts for wide opening vials
Certified microvolume inserts for wide opening vials
Inserts, with pulled point, 0,25 ml, Conical bottom, Glass, No
Catalog # HEWL5183-2085
Certified microvolume inserts for wide opening vials
Catalog # HEWL5183-2085
Certified microvolume inserts for wide opening vials
Certified microvolume inserts for wide opening vials
Inserts, with pulled point, 0,25 ml, Conical bottom, Glass, No
Price
426,00 kr
Log in for availability
1 * 100 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifications
  • Cap included:
    No
  • Capacity:
    0,25 ml
  • Description:
    Inserts, with pulled point
  • Material:
    Glass
  • Seq:
    03
  • Base:
    Conical bottom