Certified microvolume inserts for wide opening vials
Inserts, with polymer feet, graduated to 300 µl in increments of 100 µl, 0,25 ml, Conical bottom, PP, No
Catalog # HEWL5182-0549
Supplier: AGILENT HEWLETT PACKARD
Certified microvolume inserts for wide opening vials
Catalog # HEWL5182-0549
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:0,25 ml
- Description:Inserts, with polymer feet, graduated to 300 µl in increments of 100 µl
- Material:PP
- Seq:08
- Base:Conical bottom