Certified microvolume inserts for wide opening vials
Inserts, deactivated, with polymer feet, 0,25 ml, Conical bottom, Glass, No
Catalog # 548-0752
Supplier: AGILENT HEWLETT PACKARD
Certified microvolume inserts for wide opening vials
Catalog # 548-0752
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Cap included:No
- Capacity:0,25 ml
- Description:Inserts, deactivated, with polymer feet
- Material:Glass
- Seq:02
- Base:Conical bottom